missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ 24-Well, cellGrade plus-Treated, 2 in 1, 0.4 μm Pore Insert (PC)- Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
$1,426.34 - $1,565.00
Specifications
Breakapart | No |
---|---|
Bottom | Flat |
Certifications/Compliance | Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL |
Certified | Yes |
Lid | Without Lid |
Description
- Suitable for 2D and 3D cell cultures
- cellGrade™ plus surface treatment
- PC or PET membrane
- Pore sizes: 0.4µm and 8µm
- Can be used with 6-, 12-, and 24-well plates
- Sterile according to ISO 11137, SAL 10-6
- Pyrogen and endotoxin free, detection limit of 0.01 EU/mL
- Applications include: Co-culture, transmigration, chemotaxis, TEER measurements, and air-lift culture
Specifications
No | |
Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL | |
Without Lid | |
0.6 cm2 | |
Not Autoclavable | |
cellGrade™ plus | |
Yes | |
Polystyrene | |
150 to 400 μL | |
cellGrade plus | |
Yes | |
48 | |
Clear |
Flat | |
Yes | |
Ambient | |
PC | |
0.4 μm | |
Clear | |
24-well plate | |
24 | |
Sterile | |
Round | |
2D and 3D cell culture, co-culture, migration, chemotaxis, TEER assays, and air-lift culture | |
Yes | |
8.8 mm |