Please login to your online account to display your discounted pricing
Thermo Scientific™ 20mm Septa for Headspace Vials
Thermo Scientific™ National 20mm Septa for Headspace Vials are designed for use with unlined 20mm crimp seals.
$201.06 - $201.06
Specifications
Diameter (Metric) Inner 20mm
Material Aluminum Foil/White High Temp Silicone
Type Crimp Cap Septa
For Use With Headspace Vials
Item Description Aluminum Foil/White High Temp Silicone
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
03-397-46
thermo scientific™
C402037
Crimp Cap Septa Pack of 100 Each for $201.06
Description & Specifications
Specifications
Diameter (Metric) Inner 20mm
Material Aluminum Foil/White High Temp Silicone
Type Crimp Cap Septa
For Use With Headspace Vials
Item Description Aluminum Foil/White High Temp Silicone
Vial Closure Size 20mm
Thermo Scientific™ National 20mm Septa for Headspace Vials are designed for use with unlined 20mm crimp seals.
  • PTFE/red rubber septa are resistant to coring and offer good resealing characteristics
  • Gray butyl is an economical septum for low temperature applications
  • PTFE/gray butyl septum provides increased chemical resistance
  • PTFE/silicone septa offer excellent chemical resistance, resealing characteristics and low background. Use aluminum foil/silicone septa in extreme high temperature applications
  • PurePak™ PTFE/silicone septa are packed in glass jars to maintain purity up to the point of use