Learn More
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158000050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Vinylpyridine | |
100-69-6 | |
100.0 | |
Brown | |
159°C | |
Authentic | |
Glass bottle | |
1.5490 to 1.551 | |
MFCD00006355 | |
20, 256 | |
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238 | |
KGIGUEBEKRSTEW-UHFFFAOYSA-N | |
2-ethenylpyridine | |
0.1% tert.-butylcatechol | |
1.2 mPa.s (20°C) | |
97% |
97% | |
96.0 | |
-50°C | |
0.9983g/mL | |
47°C | |
96% min. (GC) | |
C7H7N | |
(C5H4N)CH=CH2 | |
5 mL | |
0.9983 | |
Solubility in water: 2.5g/100 mL in water (20°C). Other solubilities: soluble in dilute acids,soluble in hydrocarbons,alcohol,ketones and,esters,soluble in methanol,ethanol,dioxane | |
C=CC1=CC=CC=N1 | |
105.14 | |
7521 | |
105.14 | |
Liquid |
Chemical Identifiers
100-69-6 | |
105.14 | |
KGIGUEBEKRSTEW-UHFFFAOYSA-N | |
7521 | |
C=CC1=CC=CC=N1 |
C7H7N | |
MFCD00006355 | |
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238 | |
2-ethenylpyridine |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vap
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 202-879-8
RTECSNumber : UU1040000
TSCA : TSCA
RUO – Research Use Only