Please login to your online account to display your discounted pricing

2-Vinylpyrazine, 98%, stabilized, ACROS Organics™

$283.91 - $283.91

Chemical Identifiers

Chemical Name or Material 2-Vinylpyrazine
CAS 4177-16-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2
Formula Weight 106.13
MDL Number MFCD00051591
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351920050

  SDS
thermo fisher scientific
351920050
Glass bottle 5g Each for $283.91
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 106.13
Physical Form Liquid
Quantity 5g
Color Straw to dark brown
Density 1.0460g/mL
CAS Max % 100.0
Beilstein 23, V,6, 5
Refractive Index 1.5545 to 1.5575 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
CAS 4177-16-6
Molecular Formula C6H6N2
CAS Min % 97.5
Infrared Spectrum Authentic
Stabilizer Approx. 0.01% hydroquinone
Chemical Name or Material 2-Vinylpyrazine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
MDL Number MFCD00051591
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable liquid and vapor.