Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Vinylpyrazine, 98%, stab. with ca 0.1% hydroquinone

Chemical Identifiers

Chemical Name or Material 2-Vinylpyrazine
CAS 4177-16-6
Name Note stabilized with ≈0.1% hydroquinone
Molecular Formula C6H6N2
Formula Weight 106.13
MDL Number MFCD00051591
UN Number UN1993
Beilstein 105566
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1400706

  SDS
alfa aesar™
A1400706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.13
Chemical Name or Material 2-Vinylpyrazine
Name Note stabilized with ≈0.1% hydroquinone
Assay 98%
Molecular Formula C6H6N2
MDL Number MFCD00051591
CAS 4177-16-6
UN Number UN1993
Beilstein 105566
Health Hazard 1 Warning
Boiling Point 163° to 164°C
Quantity 5g
Flash Point 60°C (140°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Slightly soluble in methanol, water.

Notes
Store away from incompatible compounds such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C