missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Vinylbenzeneboronic acid, 98%

$110.69 - $344.80

Chemical Identifiers

CAS 15016-42-9
Molecular Formula C8H9BO2
Formula Weight 147.97
MDL Number MFCD01074659
Beilstein 3030430
Synonym Styrene-2-boronic acid; 2-Vinylphenylboronic acid
Chemical Name or Material 2-Vinylbenzeneboronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1982803 SDS Alfa Aesar™
L1982803
1g Each for $344.80
AAL19828MD SDS Alfa Aesar™
L19828MD
250mg Each for $110.69
Specifications

Description

Applications
2-Vinylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields. It is also used in Suzuki reaction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

15016-42-9
147.97
3030430
2-Vinylbenzeneboronic acid
C8H9BO2
MFCD01074659
Styrene-2-boronic acid; 2-Vinylphenylboronic acid

Specifications

15016-42-9
147.97
3030430
105° to 107°C
2-Vinylbenzeneboronic acid
C8H9BO2
MFCD01074659
Styrene-2-boronic acid; 2-Vinylphenylboronic acid
1g
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C