Please login to your online account to display your discounted pricing

2-Vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Vinylboronic acid pinacol ester
CAS 75927-49-0
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0% (GC)
Assay Percent Range 97+%
Molecular Formula C8H15BO2
Formula Weight 154.02
MDL Number MFCD00192492
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC395530010

  SDS
thermo fisher scientific
AC395530010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC395530050

  SDS
thermo fisher scientific
AC395530050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Low Melting Solid
Assay ≥97.0% (GC)
Formula Weight 154.02
Quantity 1g
Color White to yellow
Density 0.9080g/mL
CAS Max % 100.0
CAS Min % 97.0
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Stabilizer 500ppm (BHT) max.
CAS 75927-49-0
Chemical Name or Material Vinylboronic acid pinacol ester
Molecular Formula C8H15BO2
Assay Percent Range 97+%
MDL Number MFCD00192492
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.