Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Valerylthiophene, 97%

$27.70 - $121.15

Chemical Identifiers

Chemical Name or Material 2-Valerylthiophene
CAS 53119-25-8
Molecular Formula C9H12OS
Formula Weight 168.25
MDL Number MFCD00053149
Synonym 2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2632603

 
alfa aesar™
H2632603
1g Each for $27.70

AAH2632609

 
alfa aesar™
H2632609
10g Each for $121.15
Description & Specifications

Specifications

Formula Weight 168.25
Chemical Name or Material 2-Valerylthiophene
Assay 97%
Molecular Formula C9H12OS
MDL Number MFCD00053149
CAS 53119-25-8
Synonym 2-Pentanoylthiophene; 1-(2-Thienyl)-1-pentanone
Quantity 1g