Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Valerylfuran, 97%

$134.24 - $134.24

Chemical Identifiers

Chemical Name or Material 2-Valerylfuran
CAS 3194-17-0
Molecular Formula C9H12O2
Formula Weight 152.2
MDL Number MFCD00053147
Synonym 1-(2-Furyl)-1-pentanone; 2-Pentanoylfuran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2649109

  SDS
alfa aesar™
H2649109
10g Each for $134.24
Description & Specifications

Specifications

Formula Weight 152.2
Molecular Formula C9H12O2
CAS 3194-17-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material 2-Valerylfuran
Synonym 1-(2-Furyl)-1-pentanone; 2-Pentanoylfuran
MDL Number MFCD00053147
Quantity 10g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.