Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Triphenylphosphonio)propiophenone bromide, 98+%

$91.46 - $1,217.46

Chemical Identifiers

Chemical Name or Material 2-(Triphenylphosphonio)propiophenone bromide
CAS 2689-63-6
Molecular Formula C27H24BrOP
Formula Weight 475.37
MDL Number MFCD00050279
Beilstein 4116852
Synonym (1-Benzoylethyl)triphenylphosphonium bromide; (alpha-Methylphenacyl)triphenylphosphonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2484506

  SDS
alfa aesar™
B2484506
5g Each for $91.46

AAB2484522

  SDS
alfa aesar™
B2484522
100g Each for $1,217.46
Description & Specifications

Specifications

Formula Weight 475.37
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 4116852
Molecular Formula C27H24BrOP
Odor Odorless
Quantity 5g
CAS 2689-63-6
Synonym (1-Benzoylethyl)triphenylphosphonium bromide; (alpha-Methylphenacyl)triphenylphosphonium bromide
Chemical Name or Material 2-(Triphenylphosphonio)propiophenone bromide
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Assay ≥98%
MDL Number MFCD00050279
Melting Point 244° to 246°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.