Please login to your online account to display your discounted pricing

2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Assay Percent Range 97%
Molecular Formula C24H30ClOPSi
Formula Weight 429.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC428240010

  SDS
thermo fisher scientific
AC428240010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC428240050

  SDS
thermo fisher scientific
AC428240050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC428240250

  SDS
thermo fisher scientific
AC428240250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Quantity 1g
CAS 82495-75-8
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C24H30ClOPSi
Formula Weight 429.01