Please login to your online account to display your discounted pricing

2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride, 97%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  AC428240250

 View more versions of this product

Catalog No. AC428240250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Quantity 25g
CAS 82495-75-8
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C24H30ClOPSi
Formula Weight 429.01

Specifications

Chemical Name or Material 2-(Trimethylsilylethoxymethyl)triphenylphosphonium chloride
Assay Percent Range 97%
Molecular Formula C24H30ClOPSi
Formula Weight 429.01