Please login to your online account to display your discounted pricing

2-Trimethylsilylanisole 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Trimethylsilylanisole
CAS 704-43-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H16 O Si
Formula Weight 180.32
MDL Number MFCD09039086
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391780050

  SDS
thermo fisher scientific
AC391780050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 180.32
Quantity 5g
Color Clear colorless to slightly yellow
Density 0.9590g/mL
CAS Max % 100.0
Chemical Name or Material 2-Trimethylsilylanisole
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 704-43-8
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD09039086
CAS Min % 97.5
Molecular Formula C10 H16 O Si
Refractive Index 1.5010 to 1.5030 (20°C, 589nm)
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.