Please login to your online account to display your discounted pricing

2-(Trimethylsilyl)pyridine +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Trimethylsilyl)pyridine
CAS 13737-04-7
CAS Min % 97.0
CAS Max % 100.0
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C8H13NSi
Formula Weight 151.28g/mol
MDL Number MFCD06797181
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390360100

  SDS
thermo fisher scientific
390360100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4900 to 1.4920 (20°C, 589nm)
Density 0.911g/mL
Color Clear colorless to brown
Quantity 10g
Chemical Name or Material 2-(Trimethylsilyl)pyridine
CAS 13737-04-7
CAS Min % 97.0
CAS Max % 100.0
Physical Form Clear colorless to brown liquid
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C8H13NSi
Formula Weight 151.28g/mol
MDL Number MFCD06797181
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.