missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(Trimethylsilyl)ethynyl]aniline, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5593003

 View more versions of this product

Catalog No. AAH5593003



Specifications

Specifications

2-[(Trimethylsilyl)ethynyl]aniline
110°C (230°F)
120°C (0.1mmHg)
1g
C11H15NSi
2-2-trimethylsilyl ethynyl aniline
C[Si](C)(C)C#CC1=CC=CC=C1N
189.333
189.33
97%
0.94 g/mL
1.564
Colorless to Yellow
103529-16-4
MFCD02093646
ISXSBRZDPKQYSC-UHFFFAOYSA-N
2-(2-trimethylsilylethynyl)aniline
4377953
Liquid

Chemical Identifiers

103529-16-4
189.333
ISXSBRZDPKQYSC-UHFFFAOYSA-N
4377953
C[Si](C)(C)C#CC1=CC=CC=C1N
C11H15NSi
MFCD02093646
2-2-trimethylsilyl ethynyl aniline
2-(2-trimethylsilylethynyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents