missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(Trimethylsilyl)ethynyl]aniline, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5593014

 View more versions of this product

Catalog No. AAH5593014



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-[(Trimethylsilyl)ethynyl]aniline
Colorless to Yellow
120°C (0.1 mmHg)
C11H15NSi
MFCD02093646
2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
C[Si](C)(C)C#CC1=CC=CC=C1N
189.333
189.33
Liquid
103529-16-4
0.94 g/mL
110°C (230°F)
1.564
25 g
ISXSBRZDPKQYSC-UHFFFAOYSA-N
2-(2-trimethylsilylethynyl)aniline
4377953
97%

Chemical Identifiers

103529-16-4
189.333
ISXSBRZDPKQYSC-UHFFFAOYSA-N
4377953
C[Si](C)(C)C#CC1=CC=CC=C1N
C11H15NSi
MFCD02093646
2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
2-(2-trimethylsilylethynyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only