missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium Chloride 98.0+%, TCI America™
Supplier: TCI America T145825G
Specifications
| 2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium Chloride | |
| 165°C | |
| C24H30ClOPSi | |
| 25 g | |
| NEUMNYXEDIPGJD-UHFFFAOYSA-M | |
| triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride | |
| 11396265 | |
| ≥98.0% (T) |
| 82495-75-8 | |
| White | |
| MFCD00043159 | |
| 2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride | |
| C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-] | |
| 429.012 | |
| 429.01 | |
| Crystalline Powder |
Chemical Identifiers
| 82495-75-8 | |
| 429.012 | |
| NEUMNYXEDIPGJD-UHFFFAOYSA-M | |
| 11396265 | |
| C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-] |
| C24H30ClOPSi | |
| MFCD00043159 | |
| 2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride | |
| triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride |
Safety and Handling
TSCA : No