missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trimethylsilyl)ethanol, TCI America™

Manufacturer:  TCI America T144125ML

 View more versions of this product

Catalog No. T144125ML



Specifications

Specifications

2-(Trimethylsilyl)ethanol
Colorless
2916-68-9
MFCD00002825
2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilyl ethanol, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
C[Si](C)(C)CCO
118.251
118.25
≥96.0% (GC)
75°C
25mL
C5H14OSi
1993
ZNGINKJHQQQORD-UHFFFAOYSA-N
2-trimethylsilylethanol
18013
Clear Liquid at 20°C

Chemical Identifiers

2916-68-9
118.251
ZNGINKJHQQQORD-UHFFFAOYSA-N
18013
C[Si](C)(C)CCO
C5H14OSi
MFCD00002825
2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilyl ethanol, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
2-trimethylsilylethanol
Safety and Handling

Safety and Handling

RTECSNumber : KM5480000

TSCA : No

Documents