missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trimethylsilyl)benzothiazole, tech. 90%

$91.73 - $354.04

Chemical Identifiers

CAS 32137-73-8
Molecular Formula C10H13NSSi
Formula Weight 207.37
MDL Number MFCD00068185
Beilstein 1073964
Chemical Name or Material 2-(Trimethylsilyl)benzothiazole
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1162506 SDS Alfa Aesar™
L1162506
5g Each for $354.04
AAL1162503 SDS Alfa Aesar™
L1162503
1g Each for $91.73
Specifications Description & Specifications

Chemical Identifiers

32137-73-8
207.37
1073964
Technical Grade
C10H13NSSi
MFCD00068185
2-(Trimethylsilyl)benzothiazole

Specifications

128° to 130°C (2mmHg)
32137-73-8
207.37
1073964
90%
5g
C10H13NSSi
MFCD00068185
2-(Trimethylsilyl)benzothiazole
Technical Grade

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning