missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trimethylsilyl)benzothiazole, tech. 90%

Manufacturer:  Alfa Aesar™ L1162503

 View more versions of this product

Catalog No. AAL1162503



Specifications

Specifications

2-(Trimethylsilyl)benzothiazole
1g
C10H13NSSi
1073964
MNXBVXVIRAIAEG-UHFFFAOYSA-N
1,3-benzothiazol-2-yl(trimethyl)silane
520560
90%
128°C to 130°C (2mmHg)
32137-73-8
MFCD00068185
2-trimethylsilyl benzothiazole, 2-trimethylsilyl benzo d thiazole, 2-trimethylsilyl-1,3-benzothiazole, amtsi089, acmc-1cr1o, benzothiazole,2-trimethylsilyl, 2-trimethylsilyl benzothiazole, 2-trimethylsilyl benzothiazole, 1,3-benzothiazol-2-yl trimethyl silane, 2-trimethylsilyl-1,3-benzothiazole #
C[Si](C)(C)C1=NC2=CC=CC=C2S1
207.366
207.37
Technical

Chemical Identifiers

32137-73-8
207.366
MNXBVXVIRAIAEG-UHFFFAOYSA-N
520560
C[Si](C)(C)C1=NC2=CC=CC=C2S1
C10H13NSSi
MFCD00068185
2-trimethylsilyl benzothiazole, 2-trimethylsilyl benzo d thiazole, 2-trimethylsilyl-1,3-benzothiazole, amtsi089, acmc-1cr1o, benzothiazole,2-trimethylsilyl, 2-trimethylsilyl benzothiazole, 2-trimethylsilyl benzothiazole, 1,3-benzothiazol-2-yl trimethyl silane, 2-trimethylsilyl-1,3-benzothiazole #
1,3-benzothiazol-2-yl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents