Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Trimethylsilyl-1-ethylboronic acid pinacol ester, 97%

$106.68 - $289.30

Chemical Identifiers

Chemical Name or Material 2-Trimethylsilyl-1-ethylboronic acid pinacol ester
CAS 165904-20-1
Molecular Formula C11H25BO2Si
Formula Weight 228.22
MDL Number MFCD03788725
Beilstein 8683473
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1953403

  SDS
alfa aesar™
L1953403
1g Each for $289.30

AAL19534MD

  SDS
alfa aesar™
L19534MD
250mg Each for $106.68
Description & Specifications

Specifications

Formula Weight 228.22
Chemical Name or Material 2-Trimethylsilyl-1-ethylboronic acid pinacol ester
Molecular Formula C11H25BO2Si
Assay 97%
CAS 165904-20-1
Beilstein 8683473
Quantity 1g
MDL Number MFCD03788725