missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Trimethylsilyl-1,3-dithiane, 98+%

$431.80 - $431.80

Chemical Identifiers

CAS 13411-42-2
Molecular Formula C7H16S2Si
Molecular Weight (g/mol) 192.41
MDL Number MFCD00006655
InChI Key BTTUMVHWIAXYPJ-UHFFFAOYSA-N
Synonym 2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, 2-trimethylsilyl-1,3-dithiane, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane
PubChem CID 83413
IUPAC Name 1,3-dithian-2-yl(trimethyl)silane
SMILES C[Si](C)(C)C1SCCCS1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0097914 View Documents Alfa Aesar™
L0097914
25g Each for $431.80
 
Specifications

Chemical Identifiers

13411-42-2
192.41
BTTUMVHWIAXYPJ-UHFFFAOYSA-N
83413
C[Si](C)(C)C1SCCCS1
C7H16S2Si
MFCD00006655
2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, 2-trimethylsilyl-1,3-dithiane, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane
1,3-dithian-2-yl(trimethyl)silane

Specifications

13411-42-2
MFCD00006655
2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, 2-trimethylsilyl-1,3-dithiane, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane
C[Si](C)(C)C1SCCCS1
192.41
192.42
Stench
25g
2-Trimethylsilyl-1,3-dithiane
C7H16S2Si
1616463
BTTUMVHWIAXYPJ-UHFFFAOYSA-N
1,3-dithian-2-yl(trimethyl)silane
83413
≥98%
54°C to 55°C (0.17mmHg)
96°C (204°F)