Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trimethylsiloxy)benzaldehyde, 97%

$35.27 - $114.66

Chemical Identifiers

Chemical Name or Material 2-(Trimethylsiloxy)benzaldehyde
CAS 1078-31-5
Molecular Formula C10H14O2Si
Formula Weight 194.31
MDL Number MFCD00040721
Beilstein 1939281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1346203

  SDS
alfa aesar™
L1346203
1g Each for $35.27

AAL1346206

  SDS
alfa aesar™
L1346206
5g Each for $114.66
Description & Specifications

Specifications

Formula Weight 194.31
CAS 1078-31-5
Chemical Name or Material 2-(Trimethylsiloxy)benzaldehyde
MDL Number MFCD00040721
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 142°C (25mmHg)
Beilstein 1939281
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C10H14O2Si
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.