missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trimethylsiloxy)benzaldehyde, 97%

Manufacturer:  Alfa Aesar™ L1346206

 View more versions of this product

Catalog No. AAL1346206



Specifications

Specifications

2-(Trimethylsiloxy)benzaldehyde
5g
C10H14O2Si
1939281
GMQJTDJLZSIFQG-UHFFFAOYSA-N
2-trimethylsilyloxybenzaldehyde
66178
97%
142°C (25mmHg)
1078-31-5
MFCD00040721
2-trimethylsilyl oxy benzaldehyde, 2-trimethylsiloxy benzaldehyde, o-trimethylsilyl oxy benzaldehyde, 2-trimethylsiloxybenzaldehyde, 2-trimethylsilyl oxy benzaldehyde, benzaldehyde, 2-trimethylsilyl oxy, benzaldehyde, 2-trimethylsilyl oxy, acmc-1bs3v, 2-trimethylsilyloxybenzaldehyd, salicylaldehyde, tms derivative
C[Si](C)(C)OC1=CC=CC=C1C=O
194.305
194.31

Chemical Identifiers

1078-31-5
194.305
GMQJTDJLZSIFQG-UHFFFAOYSA-N
66178
C[Si](C)(C)OC1=CC=CC=C1C=O
C10H14O2Si
MFCD00040721
2-trimethylsilyl oxy benzaldehyde, 2-trimethylsiloxy benzaldehyde, o-trimethylsilyl oxy benzaldehyde, 2-trimethylsiloxybenzaldehyde, 2-trimethylsilyl oxy benzaldehyde, benzaldehyde, 2-trimethylsilyl oxy, benzaldehyde, 2-trimethylsilyl oxy, acmc-1bs3v, 2-trimethylsilyloxybenzaldehyd, salicylaldehyde, tms derivative
2-trimethylsilyloxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents