Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Triisopropylsilyl-1,3-dithiane, 97%

$210.34 - $820.01

Chemical Identifiers

Chemical Name or Material 2-Triisopropylsilyl-1,3-dithiane
CAS 145251-89-4
Molecular Formula C13H28S2Si
Formula Weight 276.58g/mol
MDL Number MFCD01863487
Synonym (1,3-Dithian-2-yl)triisopropylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107103

 
alfa aesar™
H51071-03
Each for $210.34

AAH5107106

 
alfa aesar™
H51071-06
Each for $820.01
Description & Specifications

Specifications

Melting Point 46° to 49°C
Quantity 1g
Chemical Name or Material 2-Triisopropylsilyl-1,3-dithiane
CAS 145251-89-4
Assay 97%
Molecular Formula C13H28S2Si
Formula Weight 276.58g/mol
MDL Number MFCD01863487
Synonym (1,3-Dithian-2-yl)triisopropylsilane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.