missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)thioxanthen-9-one, 98%

Manufacturer:  Alfa Aesar™ H6483914

 View more versions of this product

Catalog No. AAH6483914



Specifications

Specifications

2-(Trifluoromethyl)thioxanthen-9-one
Yellow
25g
C14H7F3OS
1583738
NEWRXGDGZGIHIS-UHFFFAOYSA-N
2-(trifluoromethyl)thioxanthen-9-one
74324
Crystalline
Light sensitive
147°C to 151°C
1693-28-3
MFCD00079785
2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, acmc-209dzj, pubchem10678
C1=CC=C2C(=C1)C(=O)C3=C(S2)C=CC(=C3)C(F)(F)F
280.264
280.26
98%

Chemical Identifiers

1693-28-3
280.264
NEWRXGDGZGIHIS-UHFFFAOYSA-N
74324
C1=CC=C2C(=C1)C(=O)C3=C(S2)C=CC(=C3)C(F)(F)F
C14H7F3OS
MFCD00079785
2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, acmc-209dzj, pubchem10678
2-(trifluoromethyl)thioxanthen-9-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 216-893-7

TSCA : No

Recommended Storage : Ambient temperatures

Documents