Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)thiophenol, 97%

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)thiophenol
CAS 13333-97-6
Molecular Formula C7H5F3S
Formula Weight 178.18
MDL Number MFCD00068896
UN Number UN2810
Beilstein 2441965
Synonym 2-(Trifluoromethyl)benzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1991303

  SDS
alfa aesar™
L1991303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1991306

  SDS
alfa aesar™
L1991306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.18
Chemical Name or Material 2-(Trifluoromethyl)thiophenol
CAS 13333-97-6
MDL Number MFCD00068896
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H5F3S
Synonym 2-(Trifluoromethyl)benzenethiol
Quantity 1g
Beilstein 2441965
Boiling Point 175° to 177°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
UN Number UN2810
Flash Point 68°C (154°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Assay 97%
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.