missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H54088MD

 View more versions of this product

Catalog No. AAH54088MD



Description

Description

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
250mg
C6H3F3N2O2
2-trifluoromethyl pyrimidine-5-carboxylic acid
C1=C(C=NC(=N1)C(F)(F)F)C(=O)O
192.097
192.1
170°C to 175°C
306960-77-0
MFCD09054851
RGLZXVNOHDUMRJ-UHFFFAOYSA-N
2-(trifluoromethyl)pyrimidine-5-carboxylic acid
10583889
97%

Chemical Identifiers

306960-77-0
192.097
RGLZXVNOHDUMRJ-UHFFFAOYSA-N
10583889
C1=C(C=NC(=N1)C(F)(F)F)C(=O)O
C6H3F3N2O2
MFCD09054851
2-trifluoromethyl pyrimidine-5-carboxylic acid
2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents