missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5408806

 View more versions of this product

Catalog No. AAH5408806



Description

Description

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
5g
C6H3F3N2O2
2-trifluoromethyl pyrimidine-5-carboxylic acid
RGLZXVNOHDUMRJ-UHFFFAOYSA-N
2-(trifluoromethyl)pyrimidine-5-carboxylic acid
10583889
97%
170°C to 175°C
306960-77-0
MFCD09054851
Slightly soluble in water.
C1=C(C=NC(=N1)C(F)(F)F)C(=O)O
192.097
192.1

Chemical Identifiers

306960-77-0
192.097
RGLZXVNOHDUMRJ-UHFFFAOYSA-N
10583889
C1=C(C=NC(=N1)C(F)(F)F)C(=O)O
C6H3F3N2O2
MFCD09054851
2-trifluoromethyl pyrimidine-5-carboxylic acid
2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents