Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)pyridine, 99%

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)pyridine
CAS 368-48-9
Molecular Formula C6H4F3N
Formula Weight 147.1
MDL Number MFCD02183562
UN Number UN1993
Beilstein 3112
Synonym alpha,alpha,alpha-Trifluoro-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1955403

  SDS
alfa aesar™
L1955403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.1
Chemical Name or Material 2-(Trifluoromethyl)pyridine
Molecular Formula C6H4F3N
Assay 99%
CAS 368-48-9
Beilstein 3112
Synonym alpha,alpha,alpha-Trifluoro-2-picoline
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1993
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 42°C (108°F)
Quantity 1g
Boiling Point 139° to 141°C
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
MDL Number MFCD02183562

Applications
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.

Solubility
Soluble in most organic solvents.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.