Learn More
2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
$136.36 - $547.03
Chemical Identifiers
CAS | 386704-12-7 |
---|---|
Molecular Formula | C7H4F3NO |
Molecular Weight (g/mol) | 175.11 |
MDL Number | MFCD01862647 |
InChI Key | MRPAGRCGPAXOGS-UHFFFAOYSA-N |
Synonym | 6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866 |
PubChem CID | 2777762 |
IUPAC Name | 6-(trifluoromethyl)pyridine-3-carbaldehyde |
SMILES | FC(F)(F)C1=NC=C(C=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6405403
|
Thermo Scientific Chemicals
H6405403 |
1 g |
Each for $136.36
|
|
|||||
AAH6405406
|
Thermo Scientific Chemicals
H6405406 |
5 g |
Each for $547.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
386704-12-7 | |
175.11 | |
MRPAGRCGPAXOGS-UHFFFAOYSA-N | |
2777762 | |
FC(F)(F)C1=NC=C(C=O)C=C1 |
C7H4F3NO | |
MFCD01862647 | |
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866 | |
6-(trifluoromethyl)pyridine-3-carbaldehyde |
Specifications
386704-12-7 | |
93°C (199°F) | |
MFCD01862647 | |
UN2811 | |
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866 | |
FC(F)(F)C1=NC=C(C=O)C=C1 | |
175.11 | |
175.11 | |
Solid |
52°C to 56°C | |
C7H4F3NO | |
1 g | |
Air sensitive | |
MRPAGRCGPAXOGS-UHFFFAOYSA-N | |
6-(trifluoromethyl)pyridine-3-carbaldehyde | |
2777762 | |
95% | |
2-(Trifluoromethyl)pyridine-5-carboxaldehyde |
Safety and Handling
GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H301-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only