missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%

Manufacturer:  Alfa Aesar™ H6405406

 View more versions of this product

Catalog No. AAH6405406



Specifications

Specifications

2-(Trifluoromethyl)pyridine-5-carboxaldehyde
52°C to 56°C
386704-12-7
MFCD01862647
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
C1=CC(=NC=C1C=O)C(F)(F)F
175.11
175.11
95%
Air Sensitive
5g
C7H4F3NO
UN2811
MRPAGRCGPAXOGS-UHFFFAOYSA-N
6-(trifluoromethyl)pyridine-3-carbaldehyde
2777762
Solid

Chemical Identifiers

386704-12-7
175.11
MRPAGRCGPAXOGS-UHFFFAOYSA-N
2777762
C1=CC(=NC=C1C=O)C(F)(F)F
C7H4F3NO
MFCD01862647
6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
6-(trifluoromethyl)pyridine-3-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents