Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-(Trifluoromethyl)propiophenone, 97%

$140.99 - $140.99

Chemical Identifiers

Chemical Name or Material 2'-(Trifluoromethyl)propiophenone
CAS 16185-96-9
Molecular Formula C10H9F3O
Formula Weight 202.18
MDL Number MFCD00013557
Beilstein 2263292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1771306

  SDS
alfa aesar™
A1771306
5g Each for $140.99
Description & Specifications

Specifications

Formula Weight 202.18
Chemical Name or Material 2'-(Trifluoromethyl)propiophenone
Odor Characteristic
Assay 97%
Molecular Formula C10H9F3O
MDL Number MFCD00013557
CAS 16185-96-9
Beilstein 2263292
Flash Point 101°C (213°F)
Boiling Point 220°C
Quantity 5g