missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)phenylhydrazine Hydrochloride, TCI America™

Manufacturer:  TCI America T276625G

 View more versions of this product

Catalog No. T276625G



Specifications

Specifications

2-(Trifluoromethyl)phenylhydrazine Hydrochloride
223°C
3107-34-4
MFCD00102619
ZUSWDTWYONAOPH-UHFFFAOYSA-N
[2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
12891828
Crystal-Powder at 20°C
White-Yellow
25g
C7H8ClF3N2
2-trifluoromethyl phenylhydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine hcl, 2-hydrazinobenzotrifluoride hcl, 2-hydrazinobenzotrifluoride hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine monohydrochloride, 1-2-trifluoromethyl phenyl hydrazine hydrochloride, n'-2-trifluoromethyl-phenyl-hydrazinium, chloride, 2-trifluoromethylphenylhydrazine hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride
C1=CC=C(C(=C1)C(F)(F)F)NN.Cl
212.6
212.60
≥98.0% (HPLC)

Chemical Identifiers

3107-34-4
212.6
ZUSWDTWYONAOPH-UHFFFAOYSA-N
12891828
C1=CC=C(C(=C1)C(F)(F)F)NN.Cl
C7H8ClF3N2
MFCD00102619
2-trifluoromethyl phenylhydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine hcl, 2-hydrazinobenzotrifluoride hcl, 2-hydrazinobenzotrifluoride hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine monohydrochloride, 1-2-trifluoromethyl phenyl hydrazine hydrochloride, n'-2-trifluoromethyl-phenyl-hydrazinium, chloride, 2-trifluoromethylphenylhydrazine hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride
[2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents