missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)phenylacetylene, 97%

Manufacturer:  Alfa Aesar™ H5347814

 View more versions of this product

Catalog No. AAH5347814



Description

Description

Applications
2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

2-(Trifluoromethyl)phenylacetylene
25°C (77°F)
45°C (8mmHg)
704-41-6
MFCD00467313
1-ethynyl-2-trifluoromethyl benzene, 1-ethynyl-2-trifluoromethyl-benzene, 2'-trifluoromethylphenyl acetylene, 2'-trifluoromethylphenylacetylene, 2-ethynyl-a,a,a-trifluorotoluene,1-ethynyl-2-trifluoromethylbenzene, 2-ethynyl-alpha,alpha,alpha-trifluorotoluene, 2-ethynylbenzotrifluoride, 2-trifluoromethyl phenyl ethyne, 2-trifluoromethyl phenylacetylene, o-ethynyl-trifluoromethylbenzene
OOZKONVIIMFOKW-UHFFFAOYSA-N
1-ethynyl-2-(trifluoromethyl)benzene
4100856
97%
1.222
1.476
25g
C9H5F3
UN1993
Sparingly soluble in water.
C#CC1=CC=CC=C1C(F)(F)F
170.134
170.13

Chemical Identifiers

704-41-6
170.134
OOZKONVIIMFOKW-UHFFFAOYSA-N
4100856
C#CC1=CC=CC=C1C(F)(F)F
C9H5F3
MFCD00467313
1-ethynyl-2-trifluoromethyl benzene, 1-ethynyl-2-trifluoromethyl-benzene, 2'-trifluoromethylphenyl acetylene, 2'-trifluoromethylphenylacetylene, 2-ethynyl-a,a,a-trifluorotoluene,1-ethynyl-2-trifluoromethylbenzene, 2-ethynyl-alpha,alpha,alpha-trifluorotoluene, 2-ethynylbenzotrifluoride, 2-trifluoromethyl phenyl ethyne, 2-trifluoromethyl phenylacetylene, o-ethynyl-trifluoromethylbenzene
1-ethynyl-2-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents