Learn More
2-(Trifluoromethyl)phenylacetylene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5347803
Description
2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Trifluoromethyl)phenylacetylene is used as pharmaceutical intermediate.
Solubility
Sparingly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
2-(Trifluoromethyl)phenylacetylene | |
1.222 | |
25°C (77°F) | |
1.476 | |
1 g | |
1-ethynyl-2-trifluoromethyl benzene, 2'-trifluoromethylphenyl acetylene, 2-trifluoromethyl phenylacetylene, 2-ethynyl-alpha,alpha,alpha-trifluorotoluene, 2-ethynyl-a,a,a-trifluorotoluene,1-ethynyl-2-trifluoromethylbenzene, 2'-trifluoromethylphenylacetylene, 2-ethynylbenzotrifluoride, o-ethynyl-trifluoromethylbenzene, 2-trifluoromethyl phenyl ethyne, 1-ethynyl-2-trifluoromethyl-benzene | |
OOZKONVIIMFOKW-UHFFFAOYSA-N | |
1-ethynyl-2-(trifluoromethyl)benzene | |
4100856 | |
97% |
704-41-6 | |
45°C (8 mmHg) | |
C9H5F3 | |
MFCD00467313 | |
UN1993 | |
Sparingly soluble in water. | |
C#CC1=CC=CC=C1C(F)(F)F | |
170.134 | |
170.13 |
Chemical Identifiers
704-41-6 | |
170.134 | |
OOZKONVIIMFOKW-UHFFFAOYSA-N | |
4100856 | |
C#CC1=CC=CC=C1C(F)(F)F |
C9H5F3 | |
MFCD00467313 | |
1-ethynyl-2-trifluoromethyl benzene, 2'-trifluoromethylphenyl acetylene, 2-trifluoromethyl phenylacetylene, 2-ethynyl-alpha,alpha,alpha-trifluorotoluene, 2-ethynyl-a,a,a-trifluorotoluene,1-ethynyl-2-trifluoromethylbenzene, 2'-trifluoromethylphenylacetylene, 2-ethynylbenzotrifluoride, o-ethynyl-trifluoromethylbenzene, 2-trifluoromethyl phenyl ethyne, 1-ethynyl-2-trifluoromethyl-benzene | |
1-ethynyl-2-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only