Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)phenylacetic acid, 98%

$47.32 - $47.32

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)phenylacetic acid
CAS 3038-48-0
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD00004327
Beilstein 2267339
Synonym (alpha,alpha,alpha-Trifluoro-o-tolyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2400403

  SDS
alfa aesar™
B2400403
1g Each for $47.32
Description & Specifications

Specifications

Formula Weight 204.15
Synonym (alpha,alpha,alpha-Trifluoro-o-tolyl)acetic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00004327
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C9H7F3O2
Quantity 1g
CAS 3038-48-0
Assay 98%
Melting Point 99° to 101°C
Chemical Name or Material 2-(Trifluoromethyl)phenylacetic acid
Beilstein 2267339
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.