missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl Isothiocyanate 98.0+%, TCI America™

$41.02 - $109.98

Identifiants chimiques

CAS 1743-86-8
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039644
InChI Key FCEKLQPJGXIQRY-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
PubChem CID 737162
IUPAC Name 1-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=S
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
T24591G
Afficher les documents
TCI America
T24591G
1 g
chaque for $41.02
Il en reste null
 
T24595G
Afficher les documents
TCI America
T24595G
5 g
chaque for $109.98
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

1743-86-8
203.182
FCEKLQPJGXIQRY-UHFFFAOYSA-N
737162
C1=CC=C(C(=C1)C(F)(F)F)N=C=S
C8H4F3NS
MFCD00039644
2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
1-isothiocyanato-2-(trifluoromethyl)benzene

Spécifications

1743-86-8
216°C
MFCD00039644
2810
FCEKLQPJGXIQRY-UHFFFAOYSA-N
1-isothiocyanato-2-(trifluoromethyl)benzene
737162
≥98.0% (GC)
2-(Trifluoromethyl)phenyl Isothiocyanate
Yellow
C8H4F3NS
1 g
2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
C1=CC=C(C(=C1)C(F)(F)F)N=C=S
203.182
203.18
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

FDS
missing translation for 'documents'

missing translation for 'documents'