Please login to your online account to display your discounted pricing

2-(trifluoromethyl)phenyl isothiocyanate, Acros Organics


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01593DA

 View more versions of this product

Catalog No. SB01593DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 203.19
Quantity 1g
Assay 97%
Density 1.346g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NCS
Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
CAS 1743-86-8
Molecular Formula C8H4F3NS

Specifications

Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H4F3NS
Linear Formula CF3OC6H4NCS
Formula Weight 203.19