Please login to your online account to display your discounted pricing

2-(trifluoromethyl)phenyl isothiocyanate, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H4F3NS
Linear Formula CF3OC6H4NCS
Formula Weight 203.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01593DA

  SDS
fisher scientific company
SB01593DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.19
Quantity 1g
Assay 97%
Density 1.346g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NCS
Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
CAS 1743-86-8
Molecular Formula C8H4F3NS