missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethyl)phenothiazine 98.0+%, TCI America™
Supplier: TCI America T140725G
Specifications
2-(Trifluoromethyl)phenothiazine | |
189°C | |
MFCD00005018 | |
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference | |
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F | |
267.269 | |
267.27 | |
Crystalline Powder |
92-30-8 | |
C13H8F3NS | |
25 g | |
RKGYJVASTMCSHZ-UHFFFAOYSA-N | |
2-(trifluoromethyl)-10H-phenothiazine | |
7082 | |
≥98.0% (GC) |
Chemical Identifiers
92-30-8 | |
267.269 | |
RKGYJVASTMCSHZ-UHFFFAOYSA-N | |
7082 | |
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F |
C13H8F3NS | |
MFCD00005018 | |
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference | |
2-(trifluoromethyl)-10H-phenothiazine |
Safety and Handling
EINECSNumber : (5)-1931
RTECSNumber : SP5620000
TSCA : Yes