Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)phenothiazine, 98%

$39.03 - $110.14

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)phenothiazine
CAS 92-30-8
Molecular Formula C13H8F3NS
Formula Weight 267.27
MDL Number MFCD00005018
Beilstein 226580
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2091906

  SDS
alfa aesar™
B2091906
5g Each for $39.03

AAB2091914

  SDS
alfa aesar™
B2091914
25g Each for $110.14
Description & Specifications

Specifications

Formula Weight 267.27
RTECS Number SP5620000
Assay 98%
TSCA TSCA
Molecular Formula C13H8F3NS
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
CAS 92-30-8
MDL Number MFCD00005018
Melting Point 188° to 190°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
Beilstein 226580
Chemical Name or Material 2-(Trifluoromethyl)phenothiazine
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.