missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)phenothiazine 97%, ACROS Organics™

Manufacturer:  Acros Organics AC139760050

 View more versions of this product

Catalog No. AC139760050



Specifications

Specifications

2-(Trifluoromethyl)phenothiazine
Glass bottle
187°C to 191°C
97%
96.0
97%
MFCD00005018
RKGYJVASTMCSHZ-UHFFFAOYSA-N
2-(trifluoromethyl)-10H-phenothiazine
7082
Powder
Authentic
Yellow to Green
5g
92-30-8
100.0
C13H8F3NS
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F
267.269
267.27
≥96.0%

Chemical Identifiers

92-30-8
267.269
RKGYJVASTMCSHZ-UHFFFAOYSA-N
7082
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F
C13H8F3NS
MFCD00005018
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference
2-(trifluoromethyl)-10H-phenothiazine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-145-7

RTECSNumber : SP5620000

TSCA : TSCA

SDS Documents