Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)phenothiazine 97%, ACROS Organics™

$26.86 - $26.86

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)phenothiazine
Name Note 97%
CAS 92-30-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H8F3NS
Formula Weight 267.27
MDL Number MFCD00005018
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139760050

  SDS
thermo fisher scientific
AC139760050
Glass bottle 5g Each for $26.86

AC139760250

  SDS
thermo fisher scientific
139760250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 267.27
Quantity 5g
Color Yellow to green
EINECS Number 202-145-7
Melting Point 187° to 191°C
TSCA TSCA
RTECS Number SP5620000
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Name Note 97%
Chemical Name or Material 2-(Trifluoromethyl)phenothiazine
Molecular Formula C13H8F3NS
MDL Number MFCD00005018
CAS 92-30-8
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.