Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)phenol, 98%

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)phenol
CAS 444-30-4
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00002222
UN Number UN3261
Beilstein 1867917
Synonym alpha,alpha,alpha-Trifluoro-o-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1423103

 
alfa aesar™
A1423103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1423106

 
alfa aesar™
A1423106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1423114

 
alfa aesar™
A1423114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-(Trifluoromethyl)phenol
Flash Point 65°C (149°F)
Boiling Point 147° to 148°C
Melting Point 45° to 46°C
Quantity 1g
CAS 444-30-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Odor Phenol-like
Assay 98%
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00002222
UN Number UN3261
Beilstein 1867917
Synonym alpha,alpha,alpha-Trifluoro-o-cresol
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.