missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)cyclohexanone, 97%

CAS: 56734-74-8 | C7H9F3O | 166.14 g/mol

Supplier:  Thermo Scientific Chemicals H3342303

 View more versions of this product

Catalog No. AAH3342303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(Trifluoromethyl)cyclohexanone
71°C to 73°C (12 mmHg)
MFCD00102125
UN1224
RZSQHJQIBOWYSQ-UHFFFAOYNA-N
2-(trifluoromethyl)cyclohexan-1-one
2777488
97%
56734-74-8
C7H9F3O
1 g
2-trifluoromethyl cyclohexanone, 2-trifluoromethylcyclohexanone, 2-trifluoromethyl cyclohexan-1-one, trifluoromethylcyclohexanone, 2-trifluoromethyl-cyclohexan-1-one
FC(F)(F)C1CCCCC1=O
166.14
166.14

Chemical Identifiers

56734-74-8
166.14
RZSQHJQIBOWYSQ-UHFFFAOYNA-N
2777488
FC(F)(F)C1CCCCC1=O
C7H9F3O
MFCD00102125
2-trifluoromethyl cyclohexanone, 2-trifluoromethylcyclohexanone, 2-trifluoromethyl cyclohexan-1-one, trifluoromethylcyclohexanone, 2-trifluoromethyl-cyclohexan-1-one
2-(trifluoromethyl)cyclohexan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only