missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)cinnamic Acid, TCI America™

Manufacturer:  TCI America T26085G

 View more versions of this product

Catalog No. T26085G



Specifications

Specifications

2-(Trifluoromethyl)cinnamic Acid
204°C
2062-25-1
MFCD00004381
AMVYAIXPAGBXOM-AATRIKPKSA-N
(E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid
719625
216.16
≥98.0% (GC,T)
White-Yellow
5g
C10H7F3O2
2-trifluoromethyl cinnamic acid, o-trifluoromethyl cinnamic acid, 2-trifluoromethylcinnamic acid, e-3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoromethyl phenyl prop-2-enoic acid, 3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoroformyl phenyl acrylic acid, e-2-trifluoromethyl cinnamic acid, 2-trifluoromethyl cinnamic acid, 3-2-trifluoromethyl-phenyl-acrylic acid
C1=CC=C(C(=C1)C=CC(=O)O)C(F)(F)F
216.159
CHEBI:60703
Crystal-Powder at 20°C

Chemical Identifiers

2062-25-1
216.159
AMVYAIXPAGBXOM-AATRIKPKSA-N
719625
(E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid
C10H7F3O2
MFCD00004381
2-trifluoromethyl cinnamic acid, o-trifluoromethyl cinnamic acid, 2-trifluoromethylcinnamic acid, e-3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoromethyl phenyl prop-2-enoic acid, 3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoroformyl phenyl acrylic acid, e-2-trifluoromethyl cinnamic acid, 2-trifluoromethyl cinnamic acid, 3-2-trifluoromethyl-phenyl-acrylic acid
CHEBI:60703
C1=CC=C(C(=C1)C=CC(=O)O)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents