Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Trifluoromethyl)cinnamic acid, 98+%

$26.93 - $64.65

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)cinnamic acid
CAS 2062-25-1
Molecular Formula C10H7F3O2
Formula Weight 216.16
MDL Number MFCD00004381
Beilstein 2617007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0258703

  SDS
alfa aesar™
L0258703
1g Each for $26.93

AAL0258706

  SDS
alfa aesar™
L0258706
5g Each for $64.65
Description & Specifications

Specifications

Formula Weight 216.16
Chemical Name or Material 2-(Trifluoromethyl)cinnamic acid
CAS 2062-25-1
Odor Odorless
Molecular Formula C10H7F3O2
MDL Number MFCD00004381
Beilstein 2617007
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 199° to 205°C
Quantity 1g
Assay ≥98%
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.