missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethyl)cinnamic acid, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0258706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(Trifluoromethyl)cinnamic acid | |
199°C to 205°C | |
C10H7F3O2 | |
5 g | |
2-trifluoromethyl cinnamic acid, o-trifluoromethyl cinnamic acid, 2-trifluoromethylcinnamic acid, e-3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoromethyl phenyl prop-2-enoic acid, 3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoroformyl phenyl acrylic acid, e-2-trifluoromethyl cinnamic acid, 3-2-trifluoromethyl-phenyl-acrylic acid | |
OC(=O)\C=C\C1=CC=CC=C1C(F)(F)F | |
216.16 | |
CHEBI:60703 | |
≥98% |
2062-25-1 | |
Odorless | |
MFCD00004381 | |
2617007 | |
AMVYAIXPAGBXOM-AATRIKPKSA-N | |
(2E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid | |
719625 | |
216.16 |
Chemical Identifiers
2062-25-1 | |
216.16 | |
AMVYAIXPAGBXOM-AATRIKPKSA-N | |
719625 | |
(2E)-3-[2-(trifluoromethyl)phenyl]prop-2-enoic acid |
C10H7F3O2 | |
MFCD00004381 | |
2-trifluoromethyl cinnamic acid, o-trifluoromethyl cinnamic acid, 2-trifluoromethylcinnamic acid, e-3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoromethyl phenyl prop-2-enoic acid, 3-2-trifluoromethyl phenyl acrylic acid, 2e-3-2-trifluoroformyl phenyl acrylic acid, e-2-trifluoromethyl cinnamic acid, 3-2-trifluoromethyl-phenyl-acrylic acid | |
CHEBI:60703 | |
OC(=O)\C=C\C1=CC=CC=C1C(F)(F)F |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-168-0
TSCA : No
Recommended Storage : Ambient temperatures