Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)benzylamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Name Note 98%
CAS 419308
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8F3N
Linear Formula CF3C6H4CHNH
Formula Weight 175.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC312950010

  SDS
thermo fisher scientific
AC312950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC312950050

  SDS
thermo fisher scientific
AC312950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC312950100

  SDS
thermo fisher scientific
312950100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 175.16
Quantity 1g
Color Clear colorless to light yellow
EINECS Number 221-258-2
TSCA TSCA
Density 1.249g/mL
CAS Max % 100.0
CAS 419308
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Assay Percent Range 98%
Linear Formula CF3C6H4CHNH
MDL Number MFCD00010297
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)
Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Molecular Formula C8H8F3N
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.